Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Epitizide | 1764-85-8 | 425.86 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY11196110MG

Encompass_Preferred

Epitizide is a benzothiadiazine used to produce diuresis, often combined with Triamterene. It is intended for research use only.

  • Molecular weight: 425.86
  • Formula: C10H11ClF3N3O4S3
  • CAS number: 1764-85-8
  • For research use only

Catalog No. 50-004-38968


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.