Learn More
Medchemexpress LLC Eplerenone | 107724-20-9 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0251500MG
Encompass_Preferred
Eplerenone is a selective, highly specific, and orally active aldosterone blocker (SAB). It also functions as a selective mineralocorticoid receptor antagonist (MRA) with an IC50 value of 0.081 μM. This compound is suitable for research into hypertension, atherosclerosis, chronic systolic heart failure (HF), and various cardiovascular (CV) conditions.
  • Selective, highly specific, and orally active aldosterone blocker
  • Selective mineralocorticoid receptor antagonist with an IC50 value of 0.081 μM
  • Used in research for hypertension, atherosclerosis, chronic systolic heart failure, and cardiovascular conditions
Catalog No. 50-004-05630