Learn More
Maine Molecular Quality Controls Inc ePlex RP Negative
SDP
Supplier:  Maine Molecular Quality Controls Inc M311186
Encompass
The eplex RP Negative is intended for use as an external negative quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of the pathogens listed in the cobas eplex respiratory pathogen (RP) panel performed on the cobas eplex system (Roche Diagnostics). eplex RP Negative is comprised of 6 tubes of negative control, 200 μL each.
Catalog No. NC2748698