Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eplontersen (sodium) | 2131025-75-5 | 97.0% | 8606.5 (free acid) | 1 MG
SDP

Supplier:  Medchemexpress LLC HY148089A1MG

Encompass_Preferred

Eplontersen sodium is the sodium salt of Eplontersen, a triantennary N-acetyl galactosamine (GalNAc3-7a)-conjugated antisense oligonucleotide. It targets transthyretin (TTR) mRNA, inhibiting the production of both variant and wild-type TTR protein to combat amyloid TTR (ATTR) amyloidosis diseases.

  • Targets transthyretin (TTR) mRNA
  • Inhibits variant and wild-type TTR protein production
  • Mediates N-acetylgalactosamine moiety targeting for cell delivery
  • Solid, white to off-white appearance
  • Storage as solid: -20°C, sealed, away from moisture
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-61289


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.