Learn More
Medchemexpress LLC Epoprostenol sodium | 61849-14-7 | 98.0% | 374.45 | C20H31NaO5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYA0126A10MG
Encompass_Preferred
Epoprostenol sodium is the sodium salt of prostacyclin (prostaglandin I2), supplied as a research-grade solid for studies in vascular biology and pulmonary arterial hypertension. The compound is a potent inhibitor of platelet aggregation, is supplied at high purity, and is unstable in aqueous solution so fresh solutions are recommended prior to use.
  • High purity suitable for research applications.
  • Potent inhibitor of platelet aggregation for vascular studies.
  • Supplied as a solid; store frozen to preserve stability.
  • Unstable in solution; prepare fresh aqueous solutions before experiments.
  • Suitable for studies related to pulmonary arterial hypertension and transplantation research.
Catalog No. 50-002-99218