Learn More
Medchemexpress LLC Epoprostenol sodium | 61849-14-7 | 98.03% | C20H31NaO5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYA0126A5MG
Encompass_Preferred
Epoprostenol sodium is a synthetic form of prostacyclin (prostaglandin I2) that is globally approved for treating Pulmonary arterial hypertension (PAH). It is also utilized in pulmonary hypertension and transplantation, acting as a potent inhibitor of platelet aggregation.
  • Synthetic form of prostacyclin (prostaglandin I2)
  • Approved for treating pulmonary arterial hypertension (PAH)
  • Utilized in pulmonary hypertension and transplantation
  • Functions as a potent inhibitor of platelet aggregation
  • Induces apoptosis in PAH-PASMCs
Catalog No. 50-004-32916