Learn More
Medchemexpress LLC EPOR-CD131 Heterodim 2ug
SDP
Supplier:  Medchemexpress LLC HYP730322UG
Encompass_Preferred
The EPOR Protein functions as a receptor for erythropoietin mediating erythroblast proliferation and differentiation upon EPO stimulation The heterodimer triggers the JAK2/STAT5 signaling cascade and in certain cell types activates STAT1 STAT3 and the LYN tyrosine kinase Additionally isoform EPOR-T acts as a dominant-negative receptor modulating EPOR-mediated signaling pathways EPOR-CD131 Heterodimer Protein Human (HEK293 Fc) is a recombinant protein dimer complex containing human-derived EPOR-CD131 Heterodimer protein expressed by HEK293 with C-hFc labeled tag EPOR-CD131 Heterodimer Protein Human (HEK293 Fc) has molecular weight of 110-130 kDa
Catalog No. 50-001-81032