Learn More
Medchemexpress LLC Epothilone F | 208518-52-9 | 99.5% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY127079100MG
Encompass_Preferred
Epothilone F is a 16-membered macrolide microtubule-targeting agent. It demonstrates significant anticancer activity, particularly against paclitaxel-resistant cancer cells. It inhibits the proliferation of breast cancer cells, non-small cell lung cancer cells, and drug-resistant ovarian cancer cells, and is widely used in cancer-related research.
  • Microtubule-targeting agent
  • Exhibits significant anticancer activity
  • Effective against paclitaxel-resistant cancer cells
  • Inhibits proliferation of breast cancer cells, non-small cell lung cancer cells, and drug-resistant ovarian cancer cells
  • Used in cancer-related research
  • Cytotoxicity against mouse L929 cells with an IC50 of 1.5 ng/mL
Catalog No. 50-003-40040