Learn More
Medchemexpress LLC Epothilone F | 208518-52-9 | 99.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12707910MG
Encompass_Preferred
Epothilone F is a 16-membered macrolide microtubule-targeting agent. It exhibits significant anticancer activity and is particularly effective against paclitaxel-resistant cancer cells. This compound inhibits the proliferation of breast cancer cells, non-small cell lung cancer cells, and drug-resistant ovarian cancer cells, making it widely used in cancer-related research.
  • Functions as a microtubule-targeting agent.
  • Demonstrates significant anticancer activity.
  • Effective against paclitaxel-resistant cancer cells.
  • Inhibits proliferation of breast, non-small cell lung, and drug-resistant ovarian cancer cells.
  • Widely used in cancer-related research.
  • Exhibits cytotoxicity against mouse L929 cells with an IC50 of 1.5 ng/mL.
Catalog No. 50-003-41305