Learn More
Medchemexpress LLC Epothilone F | 208518-52-9 | 99.45% | 1 MG
Supplier: Medchemexpress LLC HY1270791MG
Epothilone F is a 16-membered macrolide microtubule-targeting agent that demonstrates significant anticancer activity. It effectively inhibits paclitaxel-resistant cancer cells and has been widely applied in cancer-related research, specifically showing inhibitory effects on the proliferation of breast, non-small cell lung, and drug-resistant ovarian cancer cells.
- Potent microtubule-targeting agent.
- Exhibits significant anticancer activity.
- Effective against paclitaxel-resistant cancer cells.
- Inhibits proliferation of breast, non-small cell lung, and drug-resistant ovarian cancer cells.
- Widely used in cancer research.
- Store at -20°c, protect from light.
- Stock solutions in solvent: -80°c for 6 months; -20°c for 1 month (protect from light).