Learn More
AdipoGen Epoxidized soya bean oil, 8013-07-8, 10 g
Supplier: AdipoGen CDXE0037G010
Epoxidized soya bean oil (10 g), AdipoGen Life Sciences. Chemical. CAS: 8013-07-8. Synthetic. Useful for the determination of ESBO in foods with HPLC-MS or GC-MS. ESBO is currently employed as plasticizer and HCl scavenger in PVC products. Vegetable oils are widely used for chemical manipulation because of the high numbers of carbon-carbon double bonds. Through the epoxidation, an epoxide group (which is a more reactive group than a double bond) is added to the soybean oil, transforming it in a good hydrochloric acid scavenger and good plasticizer.