Learn More
Selleck Chemical LLC Epoxomicin 1mg 134381-21-8 BU-4061T,Aids010837
SDP
Supplier:  Selleck Chemical LLC S70381MG
Encompass_Preferred
Epoxomicin (BU-4061T, Aids010837) is a selective proteasome inhibitor with anti-inflammatory activity, inhibits primarily the CH-L activity of the 20S proteasome, while T-L and PGPH catalytic activities are also inhibited at 100- and 1000-fold reduced rate. Epoxomicin promotes apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4341