Learn More
Medchemexpress LLC Epoxyazadiradione | 18385-59-6 | 466.57 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN100965MG
Encompass_Preferred
Epoxyazadiradione is a limonoid purified from neem (Azadirachta indica) fruits. It inhibits the tautomerase activity of MIF (macrophage migration inhibitory factor) from both human (huMIF) and malaria parasites (Plasmodium falciparum (PfMIF) and Plasmodium yoelii (PyMIF)) non-competitively in a reversible fashion (Ki, 2.11-5.23 μM). It has potential against proinflammatory reactions induced by MIF of both malaria parasites and human.
  • Purified from neem (Azadirachta indica) fruits
  • Inhibits tautomerase activity of MIF from human and malaria parasites
  • Non-competitive and reversible inhibition (Ki, 2.11-5.23 μM)
  • Potential against proinflammatory reactions induced by MIF
Catalog No. 50-004-17067