Learn More
USA Scientific Inc Eppendorf Tube 5 mL, Stl
Supplier:  USA Scientific Inc 40119487
Encompass
Eppendorf Tube 5.0 mL. Conical bottom centrifuge tube with flip-open cap. Polypropylene, str, 10 bags of 20 tubes (200)
Catalog No. 50-274-4759