Learn More
USA Scientific Inc Eppendorf Tubes BioBased 15 mL
Supplier:  USA Scientific Inc 40322526
Encompass
Eppendorf Tubes BioBased, 15 mL, screw cap, str, pyrogen-, DNase-, RNase-, human and bacterial DNA-free, colorless, 500 tubes (10 bags x 50 tubes)
Catalog No. 50-274-5321