Learn More
USA Scientific Inc Eppendorf Tubes BioBased 50 mL
Supplier:  USA Scientific Inc 40322542
Encompass
Eppendorf Tubes BioBased, 50 mL, screw cap, str, pyrogen-, DNase-, RNase-, human and bacterial DNA-free, colorless, 500 tubes (20 bags x 25 tubes)
Catalog No. 50-274-5110