Learn More
Medchemexpress LLC Eprazinone dihydrochloride | 10402-53-6 | 99.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB2078A10MM/1ML
Encompass_Preferred
Eprazinone dihydrochloride is an agent with mucolytic, secretolytic, antitussive, and bronchial antispasmodic properties. It acts as a neurokinin 1 receptor (NK1R) ligand. It has potential for the treatment of chronic bronchitis by improving pulmonary function and arterial partial pressure of oxygen.
  • Acts as a neurokinin 1 receptor (NK1R) ligand.
  • Has mucolytic, secretolytic, antitussive, and bronchial antispasmodic properties.
  • Shows potential for chronic bronchitis treatment.
  • Improves pulmonary function and arterial partial pressure of oxygen.
  • Specifically displaces binding to the NK1R.
Catalog No. 50-004-09855