Learn More
Medchemexpress LLC Eprenetapopt | 5291-32-7 | 99.5% | 199.25 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1998050MG
Encompass_Preferred
Eprenetapopt (APR-246) is a first-in-class, small molecule that restores wild-type p53 functions in TP53-mutant cells. It triggers apoptosis in tumor cells and also targets the selenoprotein thioredoxin reductase 1 (TrxR1), a key regulator of cellular redox balance. Eprenetapopt acts as a p53 activator and TrxR1 inhibitor.
  • Restores wild-type p53 functions in TP53-mutant cells.
  • Triggers apoptosis in tumor cells.
  • Targets selenoprotein thioredoxin reductase 1 (TrxR1).
  • Acts as a p53 activator.
  • Acts as a TrxR1 inhibitor.
Catalog No. 50-004-04872