Learn More
Selleck Chemical LLC Pinometostat (EPZ5676) 10mg 1380288-87-8
SDP
Supplier:  Selleck Chemical LLC S706210MG
Encompass_Preferred
Pinometostat (EPZ5676) is an S-adenosyl methionine (SAM) competitive inhibitor of protein methyltransferase DOT1L with Ki of 80 pM in a cell-free assay, demonstrating >37,000-fold selectivity against all other PMTs tested, inhibits H3K79 methylation in tumor. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0536702