Learn More
TARGETMOL CHEMICALS INC EPZ004777 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T308110MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EPZ004777 is a potent selective DOT1L inhibitor with IC50 of 0.4 nM. purity: 99%
Catalog No. 50-257-7007