Learn More
Medchemexpress LLC EPZ004777 hydrochloride | 1380316-03-9 | C28H42ClN7O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15227A50MG
Encompass_Preferred
EPZ004777 hydrochloride is an effective and selective DOT1L inhibitor, reducing H3K79me2 and H3K79me1 levels and down-regulating HOXA9 and MEIS1 expression. This compound selectively inhibits the proliferation of MLL rearrangement cells, promoting their differentiation and apoptosis, making it useful for leukemia research.
  • Target: histone methyltransferase; apoptosis
  • Pathway: epigenetics; apoptosis
  • Potent and selective DOT1L inhibitor (IC50 of 0.4 nM)
  • Reduces H3K79me2 and H3K79me1 levels, down-regulating HOXA9 and MEIS1 expression
  • Selectively inhibits MLL rearrangement cells, promoting differentiation and apoptosis
  • Well tolerated in vivo with no overt toxicity observed
Catalog No. 50-003-72077