Learn More
Selleck Chemical LLC EPZ005687 5mg
SDP
Supplier:  Selleck Chemical LLC S70045MG
Encompass_Preferred
EPZ005687 is a potent and selective inhibitor of EZH2 with Ki of 24 nM in a cell-free assay, 50-fold selectivity against EZH1 and 500-fold selectivity against 15 other protein methyltransferases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3244