Learn More
Medchemexpress LLC EPZ011989 hydrochloride | 2095432-26-9 | 99.4% | 642.27 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16986B50MG
Encompass_Preferred
EPZ011989 hydrochloride is a potent and orally active Zeste Homolog 2 (EZH2) inhibitor, with a Ki value of <3 nM. It demonstrates anti-tumor activity and is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent and orally active Zeste Homolog 2 (EZH2) inhibitor
  • Shows anti-tumor activity
  • Click chemistry reagent containing an Alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-003-93563