Learn More
TARGETMOL CHEMICALS INC EPZ015666 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T607610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EPZ015666 (GSK3235025) is an orally available inhibitor of PRMT5 enzymatic activity. purity: 99%
Catalog No. 50-257-6606