Learn More
TARGETMOL CHEMICALS INC EPZ031686 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ02635MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EPZ031686 is an effective inhibitor of SMYD3 inhibitor with an IC50 of 3 nM and can be used in studies about cancer. purity: 97%
Catalog No. 50-257-8808