Learn More
Analytical Sales & Services EquaVAP 48-Well Evaporator, Efficient Sample Concentration, Stand-Alone Operation
Supplier:  Analytical Sales & Services 23048
Encompass
The EquaVAP 48-Well Evaporator is a compact benchtop solvent evaporation system designed for high-throughput applications. It efficiently evaporates common organic solvents using internal flow equalizers to ensure even gas distribution and consistent results across 48-well aluminum reaction blocks. Its modular design and step-down height adjustment make it ideal for integration into small spaces while optimizing sample concentration efficiency.
Catalog No. NC1618804