Learn More
Adam Equipment EAB 514i Equinox Analytical Balance 510g capacity x 0.0001g readability
Supplier:  Adam Equipment EAB 514I
Encompass_Preferred
Adam Equipment Equinox Analytical Balances deliver remarkable precision to labs that perform highly detailed testing. The Equinox is ideal for research and quality assurance facilities; science education; precision counting; and production and manufacturing applications needing exceptionally precise results. Eight analytical models offer capacities from 120g to 510g and 0.1mg readability. A color touch screen display provides intuitive operation, with icon-driven menus and readily accessible applications and functions. The five-inch screen is generously sized, offering 800 X 480 pixel resolution for excellent visibility. Featuring a high-resolution color touch screen, Equinox analytical balances provide intuitive operation and readily accessible functions. Equinox delivers exceptionally precise results for labs, science education and production facilities.AC adapter included.
Catalog No. NC2611847
Only null left
Add to Cart
Request a Quote