Learn More
Electron Microscopy Sciences EMS Equipment Stage 14 Place Stub
SDP
Supplier:  Electron Microscopy Sciences 450014
Encompass_Preferred
These specimen stages are all easy-change, drop-in style (no screws), and are height adjustable. 14 places for 25 or 30mm Polished embedded samples. Includes a polished Brass Tally. Compatible with all EMS series.
Catalog No. 50-190-2436