Learn More
Medchemexpress LLC HY-130800 10mg , Eragidomide CAS:1860875-51-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13080010MG
Encompass_Preferred
HY-130800 10mg Eragidomide CAS: 1860875-51-9 Eragidomide (CC-90009) is a first-in-class GSPT1-selective cereblon (CRBN) E3 ligase modulator, acts as a molecular glue. Eragidomide coopts the CRL4CRBN to selectively target GSPT1 for ubiquitination and proteasomal degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9340