Learn More
Medchemexpress LLC HY-133125 5mg Medchemexpress, ERAP1-IN-1 CAS:865273-97-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1331255MG
Encompass_Preferred
Medchemexpress, HY-133125 5mg ERAP1-IN-1 CAS:865273-97-8 ERAP1-IN-1 is an endoplasmic reticulum aminopeptidase 1 (ERAP1) inhibitor. ERAP1-IN-1 competitively inhibits ERAP1 activity towards a nonamer peptide representative of physiological substrates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1330