Learn More
Cayman Chemical Erastin, 571203-78-6, 1 mg
SDP
Supplier:  Cayman Chemical 177541
Encompass_Preferred
Molecular Formula C30H31ClN4O4, Purity ≥98%, Formula Weight 547.1; A ferroptosis inducer; induces ferroptotic cell death in vitro, an effect that can be blocked by ferroptosis inhibitors; inhibits cystine uptake by the system xc- cystine-glutamate transporter in HT-1080 fibrosarcoma and Calu-1 lung carcinoma cells at 5 M; inhibits glutamate release via the cystine-glutamate transporter in an enzyme-coupled fluorescence assay (EC50 = 1.2 M); selectively induces cell death in cells expressing the SV40 small T oncoprotein and oncogenic Ras (IC50s = 1.25-5 g/ml)
Catalog No. NC1511737