Learn More
Santa Cruz Biotechnology ErbB4/HER4 Antibody (HFR1) 200 µg/ml
Supplier:  Santa Cruz Biotechnology SC53280
Encompass
ErbB4/HER4 Antibody (HFR1) is a mouse monoclonal IgG2b κ, cited in 2 publications, provided at 200 µg/mlraised against amino acids 1249-1264 of ErbB-4 of human originrecommended for detection of ErbB-4 of mouse, rat and human origin by WB, IP, IF and IHC(P)
Catalog No. NC3243639