Learn More
Strem, An Ascensus Company CAS# 139177-64-3. 5g. Erbium(III) trifluoromethanesulfonate, min. 98% (Erbium triflate). MFCD00192126
SDP
Supplier:  Strem, An Ascensus Company 6860005G
Encompass
CAS# 139177-64-3. 5g. Erbium(III) trifluoromethanesulfonate, min. 98% (Erbium triflate). MFCD00192126. Molecular Weight: 614.47. Molecular Formula: Er(CF3SO3)3. Color/form: pink pwdr. Strem# 68-6000. http://www.strem.com/catalog/v/68-6000/
Catalog No. 50-901-12858