Learn More
Enzo Life Sciences Erbstatin Analog (5mg). CAS: 63177-57-1
SDP
Supplier:  Enzo Life Sciences BMLEI1460005
Encompass_Preferred
Erbstatin is a novel inhibitor of the EGF receptor-associated tyrosine kinase. It is an unstable compound and is completely inactivated in serum in 30 minutes. Methyl 2,5-dihydroxycinnamate is a stable erbstatin analog retaining activity after a 60 minute incubation. It inhibits EGF receptor-associated tyrosine kinase in vitro (IC50=0.77 µM). Inhibition is competitive with substrate and noncompetitive with ATP. Alternative name: Methyl 2,5-dihydroxycinnamate. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (25mg/ml), methanol or DMSO (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0567