Learn More
Cayman Chemical Erbstatin analog
SDP
Supplier:  Cayman Chemical 100102385 MG
Encompass_Preferred
A stable, potent analog of erbstatin; inhibits EGFR tyrosine kinase in vitro with an IC50 of 0.14 UG/ml; inhibits EGF-stimulated growth in NIH3T3 cells with an IC50 value of 0.5 UG/ml; delays onset of EGF-induced DNA synthesis
Catalog No. 50-288-9595