Learn More
Apexbio Technology LLC Ercalcidiol 21343-40-8 5mg
SDP
Supplier:  Apexbio Technology LLC A33955
Encompass_Preferred
Ercalcidiol (CAS 21343-40-8) is a metabolite targeting vitamin D receptor (VDR)-like receptors It is involved in modulating mineral homeostasis by participating in the physiological regulation of calcium and phosphorus balance Ercalcidiol exerts its biological activity primarily through binding to VDR-related receptors In isolated perfused rat kidney experimental models Ercalcidiol is metabolized into several di- and tri-hydroxylated vitamin D2 derivatives supporting studies of vitamin D2 metabolism pathways Based on these pharmacological properties Ercalcidiol holds research potential for assessing vitamin D nutritional status and investigating vitamin D2 metabolism
Catalog No. 50-000-95009