Learn More
Medchemexpress LLC Ercalcidiol-d3 | 1217467-39-4 | 99.9% | 415.67 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY32349S1MG
Encompass_Preferred
Ercalcidiol-d3 is the deuterium labeled Ercalcidiol, a metabolite of Vitamin D2. It can be used as an indicator of vitamin D status. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-04378