Learn More
Medchemexpress LLC Ercalcidiol-d3 (25-hydroxy vitamin D2-d3) | 1217467-39-4 | MFCD11656527 | 99.9% | 415.67 | C28H41D3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY32349S10MG
Encompass_Preferred
Ercalcidiol-d3 is the deuterium-labeled isotopologue of ercalcidiol (25-hydroxy vitamin D2), provided as a high-purity stable isotope standard for analytical and research applications. It is intended for use as a tracer or internal standard to enable accurate quantitation in studies of vitamin D metabolism and in clinical or research assays using spectrometric methods.
  • Deuterium-labeled (d3) isotopologue for isotope-dilution quantitation.
  • High purity suitable for analytical standards and reference use.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Serves as a tracer or internal standard in quantitative assays.
  • Available in multiple pack sizes, including 10 mg.
Catalog No. 50-002-96918