Learn More
Medchemexpress LLC Ercanetide (NNZ-2591) | 847952-38-9 | 100.0% | 194.23 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY148195100MG
Encompass_Preferred
Ercanetide (NNZ-2591) is a synthetic analogue of a small peptide of cyclic glycine proline (cGP) that is orally active and can cross the blood-brain barrier. It has demonstrated neuroprotective effects after ischemic brain injury and improved motor function in a rat model of Parkinson's disease.
  • Neuroprotective effects after ischemic brain injury
  • Improves motor function in a rat model of Parkinson's disease
  • Potential for research in ischemic brain injury
  • Potential for research in Angelman syndrome
Catalog No. 50-003-60583