Learn More
TARGETMOL CHEMICALS INC ERCC1-XPF-IN-2 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T609045MG
Encompass_Preferred
Also available in 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. ERCC1-XPF-IN-2 is active in nucleotide excision repair cisplatin enhancement and (gamma)H2AX assays. ERCC1-XPF-IN-2 is a potent inhibitor of ERCC1-XPF endonuclease with an IC 50 value of 0.6 uM [1]. purity: 98%
Catalog No. 50-257-9572