Learn More
Selleck Chemical LLC Erdafitinib (JNJ-42756493) 5mg 1346242-81-6
SDP
Supplier:  Selleck Chemical LLC S84015MG
Encompass_Preferred
Erdafitinib (JNJ-42756493) is a potent and selective orally bioavailable, pan fibroblast growth factor receptor (FGFR) inhibitor with potential antineoplastic activity. Erdafitinib also binds to RET (c-RET), CSF-1R, PDGFR-?/PDGFR-?, FLT4, Kit (c-Kit) and VEGFR-2 and induces cellular apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6028