Learn More
Leica Biosystems ERG.FORCEPS 7-IN HNDL 3MM 1X2
Supplier:  Leica Biosystems 38DI15545
Encompass
Available in smooth, serrated, needle tips, and with 1 x 2 teeth.The (18.5 cm) versions are for those who prefer a longer forcep and greater variety of tip choices. The curved styles are well suited for tissue separation in the water bath as well as retrieving specimens from the inside perimeter of transport jars.
Catalog No. 50-255-2415