Learn More
TARGETMOL CHEMICALS INC ERG240 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T67842
Encompass_Preferred
Also available in 1mg 5mg and bulk. Please contact Fisher for quotes. ERG240 is an inhibitor of branched-chain amino acid aminotransferase 1 (BCAT1). ERG240 can be used for the research of rheumatoid arthritis cancer and bone disease. purity: 99%
Catalog No. 50-257-6455