Learn More
Selleck Chemical LLC ERG240-E1017-100MG
SDP
Supplier:  Selleck Chemical LLC E1017100MG
Encompass_Preferred
ERG240 a selective inhibitor of branched-chain aminotransferase 1 (BCAT1) with an IC50 of 0 1-1nM leads to decreased oxygen consumption and glycolysis through the down regulation of IRG1 expression which leads to decrease itaconate production in human macrophages
Catalog No. 50-313-8531