Learn More
USA Scientific Inc ErgoOne FAST Pipette Controller
Supplier:  USA Scientific Inc 71660010
Encompass
ErgoOne FAST pipette controller. Includes wall mount holder, two 0.45 micron filters, one 0.2 micron filter, rechargeable battery, and charger.
Catalog No. NC0788915