Learn More
Medchemexpress LLC Ergosterol peroxide | 2061-64-5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN38451MG
Encompass_Preferred
Ergosterol peroxide is a steroid derivative that can be isolated from various fungi, yeast, lichens, or sponges. It exhibits anti-tumor, proapoptotic, anti-inflammatory, anti-mycobacterial, and anti-proliferative activities.
  • Isolated from natural sources such as fungi, yeast, lichens, or sponges.
  • Demonstrates anti-tumor activity.
  • Exhibits proapoptotic properties.
  • Possesses anti-inflammatory effects.
  • Shows anti-mycobacterial activity.
  • Has anti-proliferative activities.
  • High purity of 99.57%.
Catalog No. 50-004-20556