Learn More
Medchemexpress LLC HY-N1914 10mg Medchemexpress, Ergothioneine CAS:497-30-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN191410MG
Encompass_Preferred
Medchemexpress, HY-N1914 10mg Ergothioneine CAS:497-30-3 Ergothioneine, an imidazole-2-thione derivative of histidine betaine, is synthesized by certain bacteria and fungi. Ergothioneine is generally considered an antioxidant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0351