Learn More
Apexbio Technology LLC Erianin 95041-90-0 1mL(10 mM in DMSO)
SDP
Supplier:  Apexbio Technology LLC BA12235.1
Encompass_Preferred
Erianin (CAS 95041-90-0) is a naturally derived small molecule known for its antipyretic and analgesic properties Mechanistically Erianin acts by inhibiting indoleamine 2 3-dioxygenase (IDO)-mediated tumor angiogenesis thereby disrupting the immunoregulatory and pro-angiogenic pathways associated with tumor progression This compound is utilized in biomedical research to investigate mechanisms of tumor immune evasion and vascularization and to explore novel strategies for cancer therapy targeting IDO-driven angiogenic processes
Catalog No. 50-000-91396