Learn More
Medchemexpress LLC Erianin | 95041-90-0 | 318.36 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN051750MG
Encompass_Preferred
Erianin is often used as an antipyretic and analgesic agent. It could inhibit IDO-induced tumor angiogenesis. Erianin belongs to bibenzyl derivatives in structure, with activity against viruses, bacteria, and benign prostatic hyperplasia. It inhibits the growth of HeLa cells and induces apoptosis in a dose and time-dependent manner, inducing cell cycle arrest at the G2/M stage.
  • Used as an antipyretic and analgesic agent.
  • Inhibits IDO-induced tumor angiogenesis.
  • Belongs to bibenzyl derivatives.
  • Active against viruses, bacteria, and benign prostatic hyperplasia.
  • Shows significant damage to 2LL-IDO cells with low cytotoxicity to human normal liver cell line L02.
  • Inhibits the growth of cells and induces apoptosis.
  • Induces cell cycle arrest at the G2/M stage.
  • Increases expression of Bax and caspase 3, while decreasing levels of Bcl 2 and phosphorylated ERK1/2.
Catalog No. 50-003-89234