Learn More
Medchemexpress LLC Erianin | 95041-90-0 | MFCD06795132 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN051710MG
Encompass_Preferred
Erianin is a natural bibenzyl-derived small molecule reported to have antipyretic, analgesic, antibacterial, and antitumor activities. It has been studied for inhibition of IDO-induced tumor angiogenesis and is supplied for research and analytical use only. Recommended storage is 4°C with protection from light; soluble in DMSO and other formulation systems.
Catalog No. 50-002-99014